Learn More
Cole-Parmer Cole-Parmer Omega Adjustable-Volume Pipettor, 100 to 1000 L; 1/EA
Catalog No. 501960577 Shop All Cole-Parmer Products
Encompass_Preferred
These pipettors are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.
Catalog No. 50-196-0577 Supplier Cole-Parmer Supplier No. 2450108
These pipettors are offered in a wide-range of adjustable volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.