Learn More
Electron Microscopy Sciences MAGic™ Clamp Platform
SDP
Catalog No. 501928293
Encompass_Preferred
The MAGic Clamp™ provides a quick and secure way to hold Erlenmeyer flasks, tubes and microplates to platform shakers.
Catalog No. 50-192-8293 Supplier Electron Microscopy Sciences Supplier No. 8701003
The MAGic Clamp™ provides a quick and secure way to hold Erlenmeyer flasks, tubes and microplates to platform shakers.