Learn More
Medchemexpress LLC HY-18996 5mg Medchemexpress, Adjudin CAS:252025-52-8 Purity:>98%
SDP
Catalog No. 502082106
Encompass_Preferred
Medchemexpress, HY-18996 5mg Adjudin CAS:252025-52-8 Adjudin is an extensively studied male contraceptive with a superior mitochondria-inhibitory effect. Adjudin is also a potent Cl- channel blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2106 Supplier Medchemexpress LLC Supplier No. HY189965MG
Medchemexpress, HY-18996 5mg Adjudin CAS:252025-52-8 Adjudin is an extensively studied male contraceptive with a superior mitochondria-inhibitory effect. Adjudin is also a potent Cl- channel blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.