Learn More
MSC Lift-All Chain Sling, Alloy Steel, 3/8 in. Chain, 10 ft. Length, 1 ea.
Catalog No. 5000510401
Encompass
This chain sling assembly is constructed from high-strength alloy steel and features an integrated control plate and grab hook design for reliable material handling. Engineered for professional lifting applications, the assembly provides an 8,800 lb. vertical capacity and is built to rigorous industry standards.
  • Features a 10 ft. leg length with 3/8 in. grade 100 alloy steel chain
  • Designed for high visibility with yellow-finished fittings
  • Powder-coated hardware finish to resist rust and corrosion
  • Includes serialized identification on each assembly for improved traceability
  • Complies with ASME B30.9, OSHA, and NACM safety standards
  • Proof-tested and certified for dependable performance
Catalog No. 50-005-10401 Supplier MSC Supplier No. 89751747
This chain sling assembly is constructed from high-strength alloy steel and features an integrated control plate and grab hook design for reliable material handling. Engineered for professional lifting applications, the assembly provides an 8,800 lb. vertical capacity and is built to rigorous industry standards.
  • Features a 10 ft. leg length with 3/8 in. grade 100 alloy steel chain
  • Designed for high visibility with yellow-finished fittings
  • Powder-coated hardware finish to resist rust and corrosion
  • Includes serialized identification on each assembly for improved traceability
  • Complies with ASME B30.9, OSHA, and NACM safety standards
  • Proof-tested and certified for dependable performance