Learn More
MSC Lift-All Chain Sling, Control Plate & Grab Hook, 10 ft, 4300 lb Vertical Capacity
Catalog No. 5000511921
Encompass
This chain sling is designed for lifting applications, featuring a control plate and grab hook for secure and efficient handling. With a length of 10 feet and a vertical capacity of 4300 lb, this 9/32 inch alloy steel chain sling is suitable for various industrial tasks.
  • Features a grab hook and control plate sling type.
  • Abrasion-resistant for enhanced durability.
  • Meets ASME B30.9, OSHA, and NACM standards.
  • Grade 100 chain sling.
  • Powder coated finish.
  • Available in yellow and natural colors.
Catalog No. 50-005-11921 Supplier MSC Supplier No. 89751739
This chain sling is designed for lifting applications, featuring a control plate and grab hook for secure and efficient handling. With a length of 10 feet and a vertical capacity of 4300 lb, this 9/32 inch alloy steel chain sling is suitable for various industrial tasks.
  • Features a grab hook and control plate sling type.
  • Abrasion-resistant for enhanced durability.
  • Meets ASME B30.9, OSHA, and NACM standards.
  • Grade 100 chain sling.
  • Powder coated finish.
  • Available in yellow and natural colors.