Learn More
Euthanex The 512-A1-TS non-compensated single CO2 flow meter with flow rate range of 0.5 to 12 LPM.
Catalog No. NC0985379
Encompass
The 512-A1-TS non-compensated single CO2 flow meter allow customers peace of mind that the flow being displayed is accurate regardless of the variance of incoming pressure. We recommend adjustable flow meters when a variety of cage sizes are being used. Allow the user to adjust the flow based on the cages displacement size for faster results. Flow rate range is 0.5 to 12 LPM. Stand allows the user the flexibility to postion the meter in the ideal location. The TS includes a table stand, both output and input hose with quick disconnect for easy connection to our lids, and induction chambers.
Catalog No. NC0985379 Supplier Euthanex Supplier No. 512A1TS
The 512-A1-TS non-compensated single CO2 flow meter allow customers peace of mind that the flow being displayed is accurate regardless of the variance of incoming pressure. We recommend adjustable flow meters when a variety of cage sizes are being used. Allow the user to adjust the flow based on the cages displacement size for faster results. Flow rate range is 0.5 to 12 LPM. Stand allows the user the flexibility to postion the meter in the ideal location. The TS includes a table stand, both output and input hose with quick disconnect for easy connection to our lids, and induction chambers.