Learn More
Chrom Tech Inc Back Pressure Regulator Adjustable 2,000-5,000
SDP
Catalog No. NC0384601
Encompass
Biocompatible, High Pressure Adjustable Bpr Offers Flexibility To Adjust Your System Back Pressure Between 2,000 And 5, 000 Psi (138 And 345 Bar) Completely Independent Of The Flow. It'S Distinct Material Delivers Excellent Chemical Resistance And Is Specifically Designed To Be Inert To Most Solvents. It Comes With (2) F-120Blk Fittings.
Catalog No. NC0384601 Supplier Chrom Tech Inc Supplier No. P880
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Biocompatible, High Pressure Adjustable Bpr Offers Flexibility To Adjust Your System Back Pressure Between 2,000 And 5, 000 Psi (138 And 345 Bar) Completely Independent Of The Flow. It'S Distinct Material Delivers Excellent Chemical Resistance And Is Specifically Designed To Be Inert To Most Solvents. It Comes With (2) F-120Blk Fittings.