Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Adjustable Bouffant Cap
SDP

Catalog No. 19055089
Change view
Click to view available options
Color:
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-055-089 -
19-074242 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-055-089 Supplier Keystone Safety Supplier No. 110NW18BLU
This item is not returnable. View return policy

Bouffant caps keeps hair contained and are ideal for food processing, manufacturing, or general industry.

  • Polypropylene
  • 100% Latex Free
  • Economical protection
  • Manufactured in the USA

Specifications

Product Type Bouffant
Material Polypropylene
Disposable Single-use
Latex Free Yes
Model 110NW
Sterility Non-sterile
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Style Traditional
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.