Learn More
Electron Microscopy Sciences Adjustable Coverglass Rack 1 each
SDP
Catalog No. 5034077
Encompass_Preferred
This polypropylene rack accommodates a cover-glass in all sizes by simply adjusting the holder to fit the cover glass and then securing it by tightening the reset screws with the hex key. The holder is made from non ferrous material, so it is microwaveable.
The rack accommodates 10 cover glasses, round, square or rectangular, adjusting from 5 mm to 60 mm. Removable handle. Rack comes with two set of holding rods. One set accommodate cover glasses from 5 to 25 mm, and the other set for cover glasses from 5 mm to 60 mm. Rack measures 1-1/2" W x 1-1/2" H (48 x48 mm).
Catalog No. 50-340-77 Supplier Electron Microscopy Sciences Supplier No. 72243
Add to Cart
Add to Cart
This polypropylene rack accommodates a cover-glass in all sizes by simply adjusting the holder to fit the cover glass and then securing it by tightening the reset screws with the hex key. The holder is made from non ferrous material, so it is microwaveable.
The rack accommodates 10 cover glasses, round, square or rectangular, adjusting from 5 mm to 60 mm. Removable handle. Rack comes with two set of holding rods. One set accommodate cover glasses from 5 to 25 mm, and the other set for cover glasses from 5 mm to 60 mm. Rack measures 1-1/2" W x 1-1/2" H (48 x48 mm).