Learn More
Electron Microscopy Sciences Adjustable Embedding Mold, 22 x 44 x 13 mm
SDP
Catalog No. 5030348
Encompass_Preferred
These two "L's" shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Size: 22 x 44 x 13 mm
Catalog No. 50-303-48 Supplier Electron Microscopy Sciences Supplier No. 6236101
These two "L's" shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Size: 22 x 44 x 13 mm