Learn More
Electron Microscopy Sciences Adjustable Embedding Mold, 89 x146 x 25 mm
SDP
Catalog No. 5030349
Encompass_Preferred
These two "L's" shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Size: 89 x 146 x 25 mm
Catalog No. 50-303-49 Supplier Electron Microscopy Sciences Supplier No. 6236109
These two "L's" shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Size: 89 x 146 x 25 mm