Learn More
Electron Microscopy Sciences Adjustable Embedding Mold, 22 x 44 x 19 mm
SDP
Catalog No. 5030346
Encompass_Preferred
These two "L's" shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Size: 22 x 44 x 19 mm
Catalog No. 50-303-46 Supplier Electron Microscopy Sciences Supplier No. 6236103
These two "L's" shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Size: 22 x 44 x 19 mm