Learn More
Bullard™ Adjustable Flow Assemblies for RT Series Airline Respirators
SDP
Catalog No. 19807856 Shop All Bullard Company Products
Change view
Click to view available options
Connection Type:
0.25 in., Industrial Interchange Brass
0.25 in., Industrial Interchange Steel
0.25 in., Schrader Nipple
0.25 inch, industrial interchange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
19-807-856 0.25 inch, industrial interchange
19-824-393 0.25 in., Industrial Interchange Brass
19-021-472 0.25 in., Industrial Interchange Steel
19-824-395 0.25 in., Schrader Nipple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-807-856 Supplier Bullard™ Supplier No. F40
Flow control assembly for use with RT Series respirator does not include breathing tube.
  • Adjustable flow accessory
  • For use with RT series airline respirator
  • Flow control assemblies
  • Nipple types include CEJN, industrial interchange, schrader, or snap-tite in brass or steel
  • Does not include breathing tube
Specifications
Connection Type 0.25 inch, industrial interchange
Material Steel
Product Type Flow Control Valve
For Use With (Equipment) CC20™ Series Supplied Air Respirators
Includes 0.25 in. industrial interchange (Hansen compatible) quick-disconnect nipple, threaded hose connector, nylon belt
Model F40
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.