Learn More
Euthanex The 512-A1-BR non-compensated single CO2 flow meter with flow rate range of 0.5 to 12 LPM.
Catalog No. NC2099002
Encompass
The 512-A1-BR non-compensated single CO2 flow meter allow customers peace of mind that the flow being displayed is accurate regardless of the variance of incoming pressure. We recommend adjustable flow meters when a variety of cage sizes are being used. Allow the user to adjust the flow based on the cages displacement size for faster results. Flow rate range is 0.5 to 12 LPM. The wall bracket is ideal for forged turrets that are non-removable. The BR includes a wall-mounted bracket, both output and input hose with quick disconnect for easy connection to our lids, and induction chambers.
This item is not returnable. View return policy
Catalog No. NC2099002 Supplier Euthanex Supplier No. 512A1BR
The 512-A1-BR non-compensated single CO2 flow meter allow customers peace of mind that the flow being displayed is accurate regardless of the variance of incoming pressure. We recommend adjustable flow meters when a variety of cage sizes are being used. Allow the user to adjust the flow based on the cages displacement size for faster results. Flow rate range is 0.5 to 12 LPM. The wall bracket is ideal for forged turrets that are non-removable. The BR includes a wall-mounted bracket, both output and input hose with quick disconnect for easy connection to our lids, and induction chambers.