Learn More
Research Products International Corp Adjustable Freezer Rack, Holds 2 and 3 Inch Boxes, 22 1/8 x 5 1/2 x 10 Inches
SDP
Catalog No. 50476529
Encompass
This freezer storage rack provides convenient storage for both 2 and 3 inch high boxes in the same rack. Removable clips provide easy adjustment to accommodate either size box. Each rack is supplied with six L-Clips and nine T-clips. Rack is constructed of corrosion resistant stainless steel for durability. Designed for use in upright freezers. Rack accommodates up to 16 x 2 inch boxes and up to 12 x 3 inch boxes. Maximum size of box: 5.42"
Catalog No. 50-476-529 Supplier Research Products International Corp Supplier No. 247587
Add to Cart
Add to Cart
This freezer storage rack provides convenient storage for both 2 and 3 inch high boxes in the same rack. Removable clips provide easy adjustment to accommodate either size box. Each rack is supplied with six L-Clips and nine T-clips. Rack is constructed of corrosion resistant stainless steel for durability. Designed for use in upright freezers. Rack accommodates up to 16 x 2 inch boxes and up to 12 x 3 inch boxes. Maximum size of box: 5.42"