Learn More
Medicus Health Adjustable Height Ergonomic Lab Equipment Cart
SDP
Catalog No. NC2125561
Encompass
∙ Ergonomic Lab Equipment Cart has a small foot print makes it perfect for any lab, operating room, nurses station, or patient room ∙ Excellent for transporting or staging lab equipment, supplies and creating additional lab workspace ∙ Steel frame, gray laminate work top supports up to 330 lbs.Integrated, powder-coated-steel bottom shelf holds up to 55 lbs. ∙ Cart measures: 28"L x 20" W - 26"-36" Ht. ∙ Cart height adjusts to suit your needs with allen key ∙ Small foot print makes it perfect for any operating room, nurses station, or patient room ∙ Sturdy, durable cart can take the punishment of a demanding healthcare environment ∙ ADA Compliant ∙ 4" dia. caster set (2 locking, 2 non-locking) ∙ Some assembly required ∙ Cart is also available with Phenolic or ESD Protection top work surface. Call us for more details.
This item is not returnable. View return policy
Catalog No. NC2125561 Supplier Medicus Health Supplier No. 2114M2
∙ Ergonomic Lab Equipment Cart has a small foot print makes it perfect for any lab, operating room, nurses station, or patient room ∙ Excellent for transporting or staging lab equipment, supplies and creating additional lab workspace ∙ Steel frame, gray laminate work top supports up to 330 lbs.Integrated, powder-coated-steel bottom shelf holds up to 55 lbs. ∙ Cart measures: 28"L x 20" W - 26"-36" Ht. ∙ Cart height adjusts to suit your needs with allen key ∙ Small foot print makes it perfect for any operating room, nurses station, or patient room ∙ Sturdy, durable cart can take the punishment of a demanding healthcare environment ∙ ADA Compliant ∙ 4" dia. caster set (2 locking, 2 non-locking) ∙ Some assembly required ∙ Cart is also available with Phenolic or ESD Protection top work surface. Call us for more details.