Learn More
Poltex Adjustable Mouse Surgery Fixture, Magnetic Mouse Bracket
SDP
Catalog No. 22022550
Change view
Click to view available options
Animal:
Mouse, Rat
Catalog No. Animal
22-022-550 Mouse, Rat
1 options
Catalog No. 22-022-550 Supplier Poltex Supplier No. MOUSSURG
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Surgery platform: stainless steel hinges, powder coated steel work surface, magnetic platform. Folds flat for storage. Easily adjustable with rear foot in 5 degree increments from 45 - 80 degrees. Upper anesthesia tube keeper easily adjustable with thumbscrew.
  • Mouse surgery platform
  • Durable and cleanable 1/2 in. thick polypropylene and 1/4 in. thick PETG construction
  • Stainless steel hinges
  • Work surface is powder coated steel to accept magnetic anesthesia or other accessories
  • Comes with magnetic platform for patient placement
  • Unit folds flat for storage then is easily adjustable with rear foot in 5 degree increments from 45-80 degrees
  • Upper anesthesia tube keeper is easily adjustable with thumbscrew
Specifications
Autoclavable Not Autoclavable
Animal Mouse, Rat
For Use With (Application) Mouse Surgery
Type Surgical Platform
Disposable Reusable
Sterility Non-sterile