Learn More
Chemglass Life Sciences Adjustable Sampling Tube, 325mm Maximum Immersion Length, for 7L Bio Reactors
SDP
Catalog No. 501294439
Encompass
Adjustable Sampling Tube, 325mm Maximum Immersion Length, for 7L Bio Reactors. These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level. Tubing is 1/4 OD.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4439 Supplier Chemglass Life Sciences Supplier No. CLS138308
Adjustable Sampling Tube, 325mm Maximum Immersion Length, for 7L Bio Reactors. These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level. Tubing is 1/4 OD.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.