Learn More
Heidolph™ Adjustable Spanner
SDP
Catalog No. 05404078 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
StarFish™ Work Station
Catalog No. For Use With (Equipment)
05-404-078 StarFish™ Work Station
1 options
Catalog No. 05-404-078 Supplier Heidolph™ Supplier No. 015890510
Add to Cart
Add to Cart
Heidolph™ Adjustable Spanner for use with StarFish™ Work Station
  • Adjustable Spanner is used to fit the central support rod to the base plate
  • Without the use of a spanner, the assembly may unscrew and become unstable during use
Specifications
For Use With (Equipment) StarFish™ Work Station
For Use With (Application) Applies Torque to Turn Objects
Type Spanner