Learn More
Cole-Parmer Ashcroft A960AF6 Adjustable Angle Glass Thermometer, 6" Stem; 30/300 F
Catalog No. 50992737 Shop All Cole-Parmer Products
Encompass_Preferred
FEATURES
  • Mercury free blue liquid in glass with magnifying lens for easy readability
  • Molded Valox - V-shaped black case for ranges up to and including 300F
  • Die cast black aluminum V-shaped case for ranges above 300F
  • Glass front excludes dust and dirt
  • White aluminum scale with bold black graduations and figures
  • Accuracy is 0.1% of full scale
  • Easy adjustment permits case and stem to be rotated 360
  • 9" scale is standard
  • 1-1/4" - 18" NEF swivel nut
Catalog No. 50-992-737 Supplier Cole-Parmer Supplier No. 8648720
FEATURES
  • Mercury free blue liquid in glass with magnifying lens for easy readability
  • Molded Valox - V-shaped black case for ranges up to and including 300F
  • Die cast black aluminum V-shaped case for ranges above 300F
  • Glass front excludes dust and dirt
  • White aluminum scale with bold black graduations and figures
  • Accuracy is 0.1% of full scale
  • Easy adjustment permits case and stem to be rotated 360
  • 9" scale is standard
  • 1-1/4" - 18" NEF swivel nut