Learn More
Perkin Elmer US LLC Adjustable Torch Body Assembly for Avio 500
Catalog No. 502112626
Encompass_Preferred
This Torch Body Assembly is included in the Quick Change Torch Module with Cyclonic Spray Chamber/Concentric Nebulizer for Avio 500 (N0810607), Quick Change Torch Module with Scott Spray Chamber/Cross-Flow Nebulizer for Avio 500 (N0810606), and the Organics Sample Introduction System for Avio 500 (N0810609). This assembly includes: Cassette for Avio 500 Torch Body Assembly, Qty. 1 (N0810214) Viton O-Ring, 20.29 mm I.D., Qty. 2 (09902223) PTFE-Coated Blue O-Ring, 2.57 mm I.D., Qty. 2 (N0791334) Torch Knurled Nut, Qty. 1 (N0776027) Torch Washer, Qty. 1 (N0776028) Torch Spacer, Qty. 1 (N0791125) Torch Glass, Qty. 1 (N0790131)
Catalog No. 50-211-2626 Supplier Perkin Elmer US LLC Supplier No. N0810128
This Torch Body Assembly is included in the Quick Change Torch Module with Cyclonic Spray Chamber/Concentric Nebulizer for Avio 500 (N0810607), Quick Change Torch Module with Scott Spray Chamber/Cross-Flow Nebulizer for Avio 500 (N0810606), and the Organics Sample Introduction System for Avio 500 (N0810609). This assembly includes: Cassette for Avio 500 Torch Body Assembly, Qty. 1 (N0810214) Viton O-Ring, 20.29 mm I.D., Qty. 2 (09902223) PTFE-Coated Blue O-Ring, 2.57 mm I.D., Qty. 2 (N0791334) Torch Knurled Nut, Qty. 1 (N0776027) Torch Washer, Qty. 1 (N0776028) Torch Spacer, Qty. 1 (N0791125) Torch Glass, Qty. 1 (N0790131)