Learn More
MSC Adjustable Torque Wrench: 3/8" Drive, Square Drive, Foot Pound & Newton Meter
Catalog No. NC3805988
Encompass
Adjustable Torque Wrench: 3/8" Drive, Square Drive, Foot Pound & Newton Meter8.0 ft-lb Min, 36.0 ft-lb Max, 36.0 ft-lb Max, 14-11/64" OAL, Ratcheting
Catalog No. NC3805988 Supplier MSC Supplier No. 30937627
Adjustable Torque Wrench: 3/8" Drive, Square Drive, Foot Pound & Newton Meter8.0 ft-lb Min, 36.0 ft-lb Max, 36.0 ft-lb Max, 14-11/64" OAL, Ratcheting