Learn More
Electron Microscopy Sciences Adjustable Tube Rack Stand
SDP
Catalog No. 501927376
Encompass_Preferred
Tube racks may be viewed an any angle between 0 degree and 35 degree. Fits most tube racks up to 5.0" high and 8.50" wide. Simply find a comfortable angle and secure platform by tightening thumb screws. Constructed from robust 3/8" thick acrylic. Fix stand 0 degree angle to store lower profile tube racks by placing them above and below platform
Catalog No. 50-192-7376 Supplier Electron Microscopy Sciences Supplier No. 7098703
Tube racks may be viewed an any angle between 0 degree and 35 degree. Fits most tube racks up to 5.0" high and 8.50" wide. Simply find a comfortable angle and secure platform by tightening thumb screws. Constructed from robust 3/8" thick acrylic. Fix stand 0 degree angle to store lower profile tube racks by placing them above and below platform