Learn More
Med Vet International Adjustable Volume Pipettor, 0.02-0.20ml (20-200ul), J0855, 200/Pack
Catalog No. 5000582459
Encompass_Preferred
Pipettors are widely used in lab-oratories. They are an accurate method of transferring solutions or reagents. They also reduce exposure to samples or laboratory chemicals. Consisting of two parts, a reusable pipettor draws up fluid through the disposable tip which actually holds the fluid in a preset, precise volume. The fluid never reaches or touches the actual pipettor. The disposable pipettes cost only pennies and are discarded after each use. Pipettors are available in either fixed or adjustable volume. Adjustable Volume Pipettor. 200/pk.Easy volume setting by simply turning the plungerStreamlined tip for access to narrow containers and tubes
Catalog No. 50-005-82459 Supplier Med Vet International Supplier No. JOR855
Pipettors are widely used in lab-oratories. They are an accurate method of transferring solutions or reagents. They also reduce exposure to samples or laboratory chemicals. Consisting of two parts, a reusable pipettor draws up fluid through the disposable tip which actually holds the fluid in a preset, precise volume. The fluid never reaches or touches the actual pipettor. The disposable pipettes cost only pennies and are discarded after each use. Pipettors are available in either fixed or adjustable volume. Adjustable Volume Pipettor. 200/pk.Easy volume setting by simply turning the plungerStreamlined tip for access to narrow containers and tubes