Learn More
Gilson Company Inc Adjustable Wall-Mount Sieve Rack
Catalog No. NC1806214
Encompass_Preferred
Adjustable Wall-Mount Sieve Rack for 12in (305mm) diameter sieves is stainless steel with slots every 1/2in (13mm) for variable placement of the eight supplied dividers. Dividers adjust to hold eight full-height or fourteen half-height sieves. Mounts on wall, but can also be used on table, desk or counter-top.
Catalog No. NC1806214 Supplier Gilson Company Inc Supplier No. SSA822
Adjustable Wall-Mount Sieve Rack for 12in (305mm) diameter sieves is stainless steel with slots every 1/2in (13mm) for variable placement of the eight supplied dividers. Dividers adjust to hold eight full-height or fourteen half-height sieves. Mounts on wall, but can also be used on table, desk or counter-top.