Learn More
Fine Science Tools ADJUSTABLE WIRE RETRACTOR
SDP
Catalog No. NC1063433
Encompass
Product: Adjustable Wire Retractor/5cm/ Teeth: Atraumatic Blades / Teeth Length: / Teeth Width: 6mm/ Teeth Depth: 4mm/ Maximum Spread: 3.5cm/ Alloy: Stainless Steel/ Length: 5cm/ Unit: Each
Catalog No. NC1063433 Supplier Fine Science Tools Supplier No. 1700405
Product: Adjustable Wire Retractor/5cm/ Teeth: Atraumatic Blades / Teeth Length: / Teeth Width: 6mm/ Teeth Depth: 4mm/ Maximum Spread: 3.5cm/ Alloy: Stainless Steel/ Length: 5cm/ Unit: Each