Learn More
Alkali Scientific Adjustable MAGic Clamp™, for 125mL, 250mL and 500mL Flasks, 1/pk
SDP
Catalog No. 502336191
Encompass_Preferred
The MAGic Clamp™ provides a quick and secure way to hold Erlenmeyer flasks, tubes and microplates to our platform shakers. Each clamp utilizes a unique magnetic base to easily change out sizes and attachments without the use of screws and bolts. Compatible with Orbi-Shaker™ Jr orbital shaker, Orbi-Shaker™ XL orbital shaker, Orbi-Shaker™ orbital shaker, Orbi-Shaker™ CO2, Orbi-Shaker™CO2 XL, Incu-Shaker™ Mini, Incu-Shaker™ Mini CO2, Incu-Shaker™ 10L and Incu-Shaker™ 10LR.
Catalog No. 50-233-6191 Supplier Alkali Scientific Supplier No. RS70096
The MAGic Clamp™ provides a quick and secure way to hold Erlenmeyer flasks, tubes and microplates to our platform shakers. Each clamp utilizes a unique magnetic base to easily change out sizes and attachments without the use of screws and bolts. Compatible with Orbi-Shaker™ Jr orbital shaker, Orbi-Shaker™ XL orbital shaker, Orbi-Shaker™ orbital shaker, Orbi-Shaker™ CO2, Orbi-Shaker™CO2 XL, Incu-Shaker™ Mini, Incu-Shaker™ Mini CO2, Incu-Shaker™ 10L and Incu-Shaker™ 10LR.