Learn More
Kuhner Shaker Inc ADJUSTIBLE ANGLE TUBE RACK FOR
Catalog No. 502570283
Encompass
Adjustible Angle Tube Rack for 16mm (RGH-16) tubes. Holds 24 standard (TPP/Falcon) 15mL Centrifuge Tubes per rack. Compatible with all Kuhner Shaker Universal Trays.
Catalog No. 50-257-0283 Supplier Kuhner Shaker Inc Supplier No. SM317016
Adjustible Angle Tube Rack for 16mm (RGH-16) tubes. Holds 24 standard (TPP/Falcon) 15mL Centrifuge Tubes per rack. Compatible with all Kuhner Shaker Universal Trays.