Learn More
Kuhner Shaker Inc ADJUSTIBLE ANGLE TUBE RACK FOR
Supplier:  Kuhner Shaker Inc SM317018
Encompass
Adjustible Angle Tube Rack for 18mm (RGH-18) Centrifuge Tubes. Holds 24 tubes per rack. Compatible with all Kuhner Shaker Universal Trays.
Catalog No. 50-257-0282