Learn More
Cayman Chemical ADL 08-0011 (hydrochloride), Purity: ≥90%, CAS: 170098-43-8, 10 mg
SDP
Catalog No. 502950246
Encompass_Preferred
An active metabolite of alvimopan; a μ-opioid receptor antagonist (Ki = 0.25 nM); for μ-opioid receptors over κ- and δ-opioid receptors (Kis = 15.8 and 31.6 nM, respectively); inhibits endomorphin 1-induced inhibition of electrically induced contraction of isolated guinea pig ileum (pA2 = 9.4); reverses loperamide-induced delayed gastric emptying in a rat model of castor oil-induced diarrhea
Catalog No. 50-295-0246 Supplier Cayman Chemical Supplier No. 3429810 MG
An active metabolite of alvimopan; a μ-opioid receptor antagonist (Ki = 0.25 nM); for μ-opioid receptors over κ- and δ-opioid receptors (Kis = 15.8 and 31.6 nM, respectively); inhibits endomorphin 1-induced inhibition of electrically induced contraction of isolated guinea pig ileum (pA2 = 9.4); reverses loperamide-induced delayed gastric emptying in a rat model of castor oil-induced diarrhea