Learn More
Medchemexpress LLC ADL-5859 hydrochloride | 850173-95-4 | 99.0% | 50 MG
SDP
Catalog No. 5000338777
Encompass_Preferred
ADL-5859 hydrochloride is a selective and orally active δ opioid receptor (DOR) agonist. It demonstrates inhibitory activity to the hERG channel and is suitable for pain research.
  • Selective and orally active δ opioid receptor agonist
  • Reverses hyperalgesia in inflamed paw of rats
  • Shows antidepressant-like activity
  • Exhibits inhibitory activity to hERG channel
  • Inhibits cytochrome P450 2D6 (CYP2D6)
Catalog No. 50-003-38777 Supplier Medchemexpress LLC Supplier No. HY1304450MG
ADL-5859 hydrochloride is a selective and orally active δ opioid receptor (DOR) agonist. It demonstrates inhibitory activity to the hERG channel and is suitable for pain research.
  • Selective and orally active δ opioid receptor agonist
  • Reverses hyperalgesia in inflamed paw of rats
  • Shows antidepressant-like activity
  • Exhibits inhibitory activity to hERG channel
  • Inhibits cytochrome P450 2D6 (CYP2D6)