Learn More
Medchemexpress LLC Adl-5859 hydrochloride | 850173-95-4 | 99.7% | C24H29ClN2O3 | 10MG
SDP
Catalog No. 5000259787
Encompass_Preferred
ADL-5859 hydrochloride is the hydrochloride salt of a selective, orally active delta-opioid receptor agonist used as a research tool in pain pharmacology. It is supplied as a white to off-white solid with high purity and characterized potency in binding and functional assays.
  • Selective delta (δ) opioid receptor agonist with low-nanomolar affinity.
  • High purity (~99.7%) suitable for in vitro and in vivo studies.
  • Documented potency: Ki ≈ 0.84 nM, EC50 ≈ 20 nM.
  • Known off-target profile: hERG IC50 ≈ 78 μM, CYP2D6 IC50 ≈ 43 μM.
  • Supplied in small lot sizes convenient for preclinical research.
Catalog No. 50-002-59787 Supplier Medchemexpress LLC Supplier No. HY1304410MG
ADL-5859 hydrochloride is the hydrochloride salt of a selective, orally active delta-opioid receptor agonist used as a research tool in pain pharmacology. It is supplied as a white to off-white solid with high purity and characterized potency in binding and functional assays.
  • Selective delta (δ) opioid receptor agonist with low-nanomolar affinity.
  • High purity (~99.7%) suitable for in vitro and in vivo studies.
  • Documented potency: Ki ≈ 0.84 nM, EC50 ≈ 20 nM.
  • Known off-target profile: hERG IC50 ≈ 78 μM, CYP2D6 IC50 ≈ 43 μM.
  • Supplied in small lot sizes convenient for preclinical research.