Learn More
Cayman Chemical ADL5859hydrochlorIde 10mg
SDP
Catalog No. 502939710
Encompass_Preferred
A δ-opioid receptor agonist (Ki = 0.84 nM); selective for δ-opioid receptors over μ-opioid and κ-opioid receptors at 10 μM; completely reverses CFA-induced mechanical hyperalgesia in rats at 3 mg/kg; decreases immobility time in the forced swim test in rats
Catalog No. 50-293-9710 Supplier Cayman Chemical Supplier No. 2779610 MG
A δ-opioid receptor agonist (Ki = 0.84 nM); selective for δ-opioid receptors over μ-opioid and κ-opioid receptors at 10 μM; completely reverses CFA-induced mechanical hyperalgesia in rats at 3 mg/kg; decreases immobility time in the forced swim test in rats