Learn More
Enzo Life Sciences NG,N'G-Dimethyl-L-arginine . dihydrochloride (25mg)
SDP
Supplier:  Enzo Life Sciences ALX106008M025
Encompass_Preferred
Does not inhibit nitric oxide synthesis, both in vitro and in vivo. Alternative name: SDMA . 2HCl, H-Arg(di-Me)-OH . 2HCl (symmetrical). Formula: C8H18N4O2 . 2HCl. MW: 202.2 . 73.0. Purity: ≥98% (TLC). Appearance: White to off-white powder. Solubility: Soluble in water (100mg/ml). Long Term Storage: -20°C. Handling: Hygroscopic.
Catalog No. 50-200-7738