Learn More
Enzo Life Sciences NG,NG-Dimethyl-L-arginine . dihydrochloride (25mg). CAS: 220805-22-1
SDP
Supplier:  Enzo Life Sciences ALX106006M025
Encompass_Preferred
Endogenous nitric oxide synthase (NOS) inhibitor. Alternative name: H-Arg(di-Me)-OH . 2HCl (asymmetrical), ADMA . 2HCl. Formula: C8H18N4O2 . 2 HCl . H2O. MW: 202.2 . 73.0 . 18.0. Purity: ≥97% (TLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-7736