Learn More
Enzo Life Sciences ADMA (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850323KI01
Encompass_Preferred
The ADMA (human) ELISA kit is a complete assay for the quantitative determination of ADMA concentration. This kit is to be used with serum, citrate- and EDTA-plasma from human origin. The ADMA (human) ELISA has no cross-reactivity with SDMA, NMMA or L-arginine. Assay Time: 15 to 20 hours + 1 hour. Applications: ELISA . Species reactivity: Human .
Catalog No. 50-201-0161