Learn More
Med Vet International Primary Blood Administration Set With Clave Y-Sites And Check Valve, 100", 48Ea/Case
Catalog No. 502830601
Encompass_Preferred
Primary Blood Administration Set with Clave Y-Sites and Check Valve, 100"Set is supplied with two nonvented piercing pins, CAIR clamp, slide clamp, drip chamber with 200 micron blood filter It also includes two prepierced Y-sites, secure lock male adapter and blood bulb pump
Catalog No. 50-283-0601 Supplier Med Vet International Supplier No. ABB1266328
Primary Blood Administration Set with Clave Y-Sites and Check Valve, 100"Set is supplied with two nonvented piercing pins, CAIR clamp, slide clamp, drip chamber with 200 micron blood filter It also includes two prepierced Y-sites, secure lock male adapter and blood bulb pump