Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Admittance Signs

Catalog No. 19807064 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Aluminum
Fiberglass, Polyester
Plastic
Color:
Blue on White
Red on White
Height (English):
7 in.
10 in.
Displayed Text:
AUTHORIZED PERSONNEL ONLY
Notice No Entry Unless Authorized
Restricted Area Authorized Employees Only
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Color Height (English) Displayed Text
19-807-064 Aluminum Blue on White 10 in. Notice No Entry Unless Authorized
17-046-6 Fiberglass, Polyester Red on White 10 in. AUTHORIZED PERSONNEL ONLY
19-016593 Fiberglass, Polyester Red on White 10 in. Restricted Area Authorized Employees Only
19-017711 Fiberglass, Polyester Blue on White 7 in. AUTHORIZED PERSONNEL ONLY
19-103-667 Plastic Red on White 10 in. Restricted Area Authorized Employees Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-807-064 Supplier Brady™ Supplier No. 43479
  • $59.50
    $36.65 / Each of 1

    Save $22.85

    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alert employees to potential hazards, safety warning and cautionary notices

  • Indicate and identify areas of access
  • Restrict access to property
  • Available in Fiberglass reinforced polyester, aluminum, plastic and polyester
  • Available in variety of sizes

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145, ASTM D 1593
Material Aluminum
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Use in industrial, utility, commercial and institutional environments, common uses include safety signs, utility signs, information signs and equipment warning signs, recommended for both indoor and outdoor use
Height (English) 10 in.
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.04 in.
Width (English) 14 in.
Temperature Range (Metric) -40°C to 110°C
Shape Rectangular
Temperature Range (English) -40°F to 230°F
Languages English
Displayed Text Notice No Entry Unless Authorized
Product Type Admittance Sign
Color Blue on White
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.