Learn More
Aviva Systems Biology Corp ADNP Antibody - N-terminal region (ARP39186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADNP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADNP. Alternate names: ADNP1, KIAA0784ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56935