Learn More
Cayman Chemical Adomeglivant, Purity: ≥98%, CAS: 1488363-78-5, 10 mg
SDP
Catalog No. 502978748
Encompass_Preferred
An allosteric antagonist of the glucagon receptor; inhibits glucagon-induced increases in cAMP levels in HEK293 cells expressing the rat glucagon or GLP-1 receptors (IC50s = 1.8 and 1.2 µM, respectively) but has no effect on cAMP levels when used alone; inhibits increases in cAMP levels induced by GLP-1 or exendin-4 in HEK293 cells expressing the GLP-1 receptor (IC50s = 7 and 12 µM, respectively); decreases glucose-stimulated insulin secretion at 1 µM in combination with exendin (9-39) in isolated human islets but not glucose-stimulated glucagon secretion alone or in combination with exendin (9-39) in isolated mouse islets
Catalog No. 50-297-8748 Supplier Cayman Chemical Supplier No. 2966810 MG
An allosteric antagonist of the glucagon receptor; inhibits glucagon-induced increases in cAMP levels in HEK293 cells expressing the rat glucagon or GLP-1 receptors (IC50s = 1.8 and 1.2 µM, respectively) but has no effect on cAMP levels when used alone; inhibits increases in cAMP levels induced by GLP-1 or exendin-4 in HEK293 cells expressing the GLP-1 receptor (IC50s = 7 and 12 µM, respectively); decreases glucose-stimulated insulin secretion at 1 µM in combination with exendin (9-39) in isolated human islets but not glucose-stimulated glucagon secretion alone or in combination with exendin (9-39) in isolated mouse islets