Learn More
Research Products International Corp Adonitol, 500 G
SDP
Catalog No. 501976981
Encompass_Preferred
Adonitol is a crystalline pentose alcohol formed by the reduction of ribose. It occurs naturally in the plant Adonis vernalis, as well as in the cell walls of Gram positive bacteria.
Catalog No. 50-197-6981 Supplier Research Products International Corp Supplier No. A32000500.0
Adonitol is a crystalline pentose alcohol formed by the reduction of ribose. It occurs naturally in the plant Adonis vernalis, as well as in the cell walls of Gram positive bacteria.