Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adooq Bioscience LLC ADOOQ BIOSCIENCE LLC Setmelanotide, Selective MC4R Agonist, CAS 920014-72-8

Catalog No. NC3994607
Encompass

Setmelanotide is a selective melanocortin 4 receptor (MC4R) agonist, effective at EC50s of 0.27 nM for human MC4R and 0.28 nM for rat MC4R. This compound is for research use only.

  • Selective melanocortin 4 receptor (MC4R) agonist
  • EC50s of 0.27 nM for human MC4R
  • EC50s of 0.28 nM for rat MC4R
  • Formula: C49H68N18O9S2
  • Molecular weight: 1117.31
  • CAS number: 920014-72-8
  • Store lyophilized at -20°C, keep desiccated
  • Lyophilized form stable for 36 months
  • In solution, store at -20°C and use within 1 month to prevent loss of potency
  • Aliquot to avoid multiple freeze/thaw cycles
  • For research use only

Catalog No. NC3994607 Supplier Adooq Bioscience LLC Supplier No. A206895MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Setmelanotide is a selective melanocortin 4 receptor (MC4R) agonist, effective at EC50s of 0.27 nM for human MC4R and 0.28 nM for rat MC4R. This compound is for research use only.

  • Selective melanocortin 4 receptor (MC4R) agonist
  • EC50s of 0.27 nM for human MC4R
  • EC50s of 0.28 nM for rat MC4R
  • Formula: C49H68N18O9S2
  • Molecular weight: 1117.31
  • CAS number: 920014-72-8
  • Store lyophilized at -20°C, keep desiccated
  • Lyophilized form stable for 36 months
  • In solution, store at -20°C and use within 1 month to prevent loss of potency
  • Aliquot to avoid multiple freeze/thaw cycles
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.