Learn More
Aviva Systems Biology Corp ADORA2A Antibody - N-terminal region (ARP59952_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADORA2A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ADORA2A. Alternate names: ADORA2, RDC8, hA2aRApplicationWB. Species reactivity: Dog, Human, Rabbit, Rat
Catalog No. 50-286-892