Learn More
Eurofins Discoverx ChemiBrite A2A Adenosine Receptor Stable Cell Line, 2 vials. (HTS048L)
Supplier:  Eurofins Discoverx HTS048L
Encompass
The ChemiBrite A2A Adenosine Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of ADORA2A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADORA2A alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of ADORA2A, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5750
Add to Cart
Request a Quote