Learn More
Eurofins Discoverx Ready-to-Assay A2A Adenosine Receptor Frozen Cells, 2 Vials of Cells. (HTS048LRTA)
Catalog No. 502005751
Encompass
The Ready-to-Assay A2A Adenosine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. adenosine, NECA) based activation of ADORA2A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADORA2A alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of ADORA2A, the resulting calcium activity can be assayed in either detection mode. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5751 Supplier Eurofins Discoverx Supplier No. HTS048LRTA
The Ready-to-Assay A2A Adenosine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. adenosine, NECA) based activation of ADORA2A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADORA2A alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of ADORA2A, the resulting calcium activity can be assayed in either detection mode. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.