Learn More
ABclonal Technology ADORA2A Rabbit pAb
SDP
Catalog No. 5000105040
Encompass_Preferred
This gene encodes a member of the G protein-coupled receptor (GPCR) superfamily, specifically an A2A adenosine receptor. It responds to adenosine, activating G(s) and G(olf) proteins to increase intracellular cAMP levels. The receptor plays key roles in regulating cardiac rhythm, circulation, cerebral and renal blood flow, immune function, pain, and sleep. It is involved in conditions like inflammatory diseases and neurodegenerative disorders. Alternative splicing produces multiple transcript variants, including a read-through transcript with the upstream SPECC1L gene, thought to be non-coding.
Catalog No. 50-001-05040 Supplier ABclonal Technology Supplier No. A1587500UL
This gene encodes a member of the G protein-coupled receptor (GPCR) superfamily, specifically an A2A adenosine receptor. It responds to adenosine, activating G(s) and G(olf) proteins to increase intracellular cAMP levels. The receptor plays key roles in regulating cardiac rhythm, circulation, cerebral and renal blood flow, immune function, pain, and sleep. It is involved in conditions like inflammatory diseases and neurodegenerative disorders. Alternative splicing produces multiple transcript variants, including a read-through transcript with the upstream SPECC1L gene, thought to be non-coding.