Learn More
Selleck Chemical LLC ADP-25mg
SDP
Supplier:  Selleck Chemical LLC S936825MG
Encompass_Preferred
ADP (Adenosine diphosphate, adenosine pyrophosphate, Adenosine 5'-diphosphate) is an important organic compound in metabolism and is essential to the flow of energy in living cells.
Catalog No. 50-238-9739