Learn More
Chrono Log Corporation ADP
SDP
Catalog No. NC9660687
Encompass
This item has a minimum qty of 2 per supplier requirements.
ADP Aggregation Reagent, Each vial contains 2.5 mg of lyophilized adenosine diphosphate, Performs 500 to 1, 000 tests
Catalog No. NC9660687 Supplier Chrono Log Corporation Supplier No. 384
This item has a minimum qty of 2 per supplier requirements.
ADP Aggregation Reagent, Each vial contains 2.5 mg of lyophilized adenosine diphosphate, Performs 500 to 1, 000 tests