Learn More
Sigma Aldrich Fine Chemicals Biosciences ADP/ATP Ratio Assay Kit su
Catalog No. 502904268
Encompass
Changes in the ADP/ATP ratio have been used to differentiate modes of cell death and viability. Increased levels of ATP and decreased levels of ADP signify proliferating cells. Conversely decreased levels of ATP and increased levels of ADP represent apoptotic or necrotic cells where the decrease in ATP and increase in ADP are much more pronounced in necrosis versus apoptosis.
Catalog No. 50-290-4268 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1351KT
Changes in the ADP/ATP ratio have been used to differentiate modes of cell death and viability. Increased levels of ATP and decreased levels of ADP signify proliferating cells. Conversely decreased levels of ATP and increased levels of ADP represent apoptotic or necrotic cells where the decrease in ATP and increase in ADP are much more pronounced in necrosis versus apoptosis.