Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ADP-Glucose disodium | 102129-65-7 | 95.0% | 633.31 | 25 MG
SDP

Catalog No. 5000348040
Encompass_Preferred

ADP-Glucose disodium is an immediate precursor for the biosynthesis of storage polysaccharides in plants, green algae, and cyanobacteria, and structural polysaccharides in some bacteria, through the addition of glucose. It is utilized to produce amylose, amylopectin, starch, and other polysaccharides by amylose synthase or starch synthase in plastids. ADPG is typically produced in plastids, though it can also be biosynthesized in the cytoplasm of certain grasses and imported into plastids by membrane-bound transporters.

  • Precursor for polysaccharide biosynthesis
  • Utilized in production of amylose, amylopectin, and starch
  • Soluble in H2O at 35 mg/mL or greater
  • Recommended storage at -20°C, sealed, away from moisture
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-48040 Supplier Medchemexpress LLC Supplier No. 25MGHY136830
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ADP-Glucose disodium is an immediate precursor for the biosynthesis of storage polysaccharides in plants, green algae, and cyanobacteria, and structural polysaccharides in some bacteria, through the addition of glucose. It is utilized to produce amylose, amylopectin, starch, and other polysaccharides by amylose synthase or starch synthase in plastids. ADPG is typically produced in plastids, though it can also be biosynthesized in the cytoplasm of certain grasses and imported into plastids by membrane-bound transporters.

  • Precursor for polysaccharide biosynthesis
  • Utilized in production of amylose, amylopectin, and starch
  • Soluble in H2O at 35 mg/mL or greater
  • Recommended storage at -20°C, sealed, away from moisture
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.